CAS RN 1253641-65-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ TM 38837
Highly potent CB1 inverse agonist; peripherally restricted