CAS RN 1254944-66-5
CAS RN 1254944-66-5
IUPAC Name:
ethyl 2-[5-(methylsulfanyl)thiophene-2-carbonyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylate
Synonyms:
ethyl 2-[5-(methylsulfanyl)thiophene-2-carbonyl]-3,4-dihydro-1H-isoquinoline-3-carboxylate
Molecular Weight (g/mol):
361.47
Molecular Formula:
C18H19NO3S2
InChi Key:
UIEBLUZPSFAFOC-UHFFFAOYNA-N
SMILES:
CCOC(=O)C1CC2=CC=CC=C2CN1C(=O)C1=CC=C(SC)S1