accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNOHO

CAS RN 1268882-43-4

IUPAC Name: 8,9-diazatricyclo[4.3.0.0²,⁴]nona-1(6),7-diene-7-carboxylic acid
Synonyms: 8,9-diazatricyclo[4.3.0.0²,⁴]nona-1(6),7-diene-7-carboxylic acid
Molecular Weight (g/mol): 164.16
Molecular Formula: C8H8N2O2
InChi Key: CUTZNERBKDMLAP-UHFFFAOYNA-N
SMILES: OC(=O)C1=NNC2=C1CC1CC21
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results