accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBOHHO

CAS RN 127972-02-5

IUPAC Name: (5-formyl-2-methoxyphenyl)boronic acid
Synonyms: 5-formyl-2-methoxyphenylboronic acid
Molecular Weight (g/mol): 179.97
Molecular Formula: C8H9BO4
InChi Key: NKKNXLPHCRLBDY-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C=C1B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)