CAS RN 128363-76-8
CAS RN 128363-76-8
IUPAC Name:
di-tert-butylphosphane borane
Synonyms:
di-tert-butylphosphane borane
Molecular Weight (g/mol):
160.05
Molecular Formula:
C8H22BP
InChi Key:
JPINHYFOWCBINQ-UHFFFAOYSA-N
SMILES:
B.CC(C)(C)PC(C)(C)C
1
–
2
of
2
results
1
Thermo Scientific Chemicals Borane-di(tert-butyl)phosphine complex, 94%, Thermo Scientific™
CAS: 128363-76-8 Molecular Formula: C8H22BP Molecular Weight (g/mol): 160.05 MDL Number: MFCD03412071 InChI Key: JPINHYFOWCBINQ-UHFFFAOYSA-N Synonym: di-tert-butylphosphane boranylidyne,borane di tert-butyl phosphine complex,borane-di tert-butyl phosphine complex,di-t-butylphosphine-borane,di-tert-butylphosphine borane,di-tert-butylphosphane borane,di-tert-butylphosphine trihydroboron,borane di-tert-butylphosphine complex,boron-di-tert-butylphosphane 1/1 PubChem CID: 22572032 IUPAC Name: di-tert-butylphosphane borane SMILES: B.CC(C)(C)PC(C)(C)C