accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHN

CAS RN 129-56-6

IUPAC Name: 14,15-diazatetracyclo[7.6.1.0²,⁷.0¹³,¹⁶]hexadeca-1(15),2,4,6,9(16),10,12-heptaen-8-one
Synonyms: 1,9-pyrazoloanthrone
Molecular Weight (g/mol): 220.23
Molecular Formula: C14H8N2O
InChi Key: ACPOUJIDANTYHO-UHFFFAOYSA-N
SMILES: O=C1C2=C3C(NN=C3C3=CC=CC=C13)=CC=C2
Product Type 
  • (3)
  • (2)
Form 
  • (3)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results