CAS RN 130964-39-5
IUPAC Name:
N-(2-{[3-(4-bromophenyl)prop-2-en-1-yl]amino}ethyl)isoquinoline-5-sulfonamide hydrate dihydrochloride
Synonyms:
N-(2-{[3-(4-bromophenyl)prop-2-en-1-yl]amino}ethyl)isoquinoline-5-sulfonamide hydrate dihydrochloride
Molecular Weight (g/mol):
537.29
Molecular Formula:
C20H24BrCl2N3O3S
InChi Key:
GKFFJFGBWAGAFD-UHFFFAOYSA-N
SMILES:
O.Cl.Cl.BrC1=CC=C(C=CCNCCNS(=O)(=O)C2=C3C=CN=CC3=CC=C2)C=C1
1
–
1
of
1
results
1
R&D Systems™ H 89 dihydrochloride
CAS: 130964-39-5 Molecular Formula: C20H24BrCl2N3O3S