accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNOONNO

CAS RN 131631-89-5

IUPAC Name: N-(3-{4-[4-(2-oxo-1,2,3,4-tetrahydroquinolin-1-yl)piperidine-1-carbonyl]phenoxy}propyl)acetamide
Synonyms: N-(3-{4-[4-(2-oxo-3,4-dihydroquinolin-1-yl)piperidine-1-carbonyl]phenoxy}propyl)acetamide
Molecular Weight (g/mol): 449.55
Molecular Formula: C26H31N3O4
InChi Key: KSNUCNRMDYJBKT-UHFFFAOYSA-N
SMILES: CC(=O)NCCCOC1=CC=C(C=C1)C(=O)N1CCC(CC1)N1C(=O)CCC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results