Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ OPC 21268
GREENER_CHOICESDP

Catalog No. 392450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-245-0 50 mg
39-241-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-245-0 Supplier R&D Systems™ Supplier No. 3924/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Orally active vasopressin V1 antagonist

Nonpeptide vasopressin V1 receptor antagonist. Displays greater affinity for rat V1 than human V1 (Ki values are 25 and 8800 nM respectively). Exhibits 1000-fold selectivity for V1 receptors over V2 receptors. Orally active.

Chemical Identifiers

CAS 131631-89-5
Molecular Formula C26H31N3O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 449.54
Purity >98%
Molecular Weight (g/mol) 449.551
Chemical Name or Material OPC 21268
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.