accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClONHFFFOOH

CAS RN 1322598-09-3

IUPAC Name: 3-[2-(3-chlorophenyl)acetamido]-4-[4-(trifluoromethyl)phenyl]butanoic acid
Synonyms: 3-[2-(3-chlorophenyl)acetamido]-4-[4-(trifluoromethyl)phenyl]butanoic acid
Molecular Weight (g/mol): 399.79
Molecular Formula: C19H17ClF3NO3
InChi Key: QOSIJVVNNGXEKE-UHFFFAOYNA-N
SMILES: OC(=O)CC(CC1=CC=C(C=C1)C(F)(F)F)NC(=O)CC1=CC(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)