accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3C(S)HO(R)NHO(R)H3C(S)HOOS(R)HNCH3OOHO

CAS RN 133343-34-7

IUPAC Name: (2R)-2-acetamido-3-[(2R,3S,4R)-3-hydroxy-2-[(1S)-1-hydroxy-3-methylbutyl]-4-methyl-5-oxopyrrolidine-2-carbonylsulfanyl]propanoic acid
Synonyms: lactacystin
Molecular Weight (g/mol): 390.45
Molecular Formula: C16H26N2O7S
InChi Key: KZZNIEYFCKAJPQ-VRKAVTRVSA-N
SMILES: CC(C)C[C@H](O)[C@]1(NC(=O)[C@H](C)[C@@H]1O)C(=O)SC[C@H](NC(C)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results