accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 134030-71-0

BONNSONNSONNSTl

CAS RN 134030-71-0

IUPAC Name: λ¹-thallanylium tris({[3-(thiophen-2-yl)-1H-pyrazol-1-yl]oxy})-λ²-boranuide
Synonyms: λ¹-thallanylium tris({[3-(thiophen-2-yl)pyrazol-1-yl]oxy})-λ²-boranuide
Molecular Weight (g/mol): 710.76
Molecular Formula: C21H15BN6O3S3Tl
InChi Key: CLSWVTDHYMQQPF-UHFFFAOYSA-N
SMILES: [Tl+].O([B-](ON1C=CC(=N1)C1=CC=CS1)ON1C=CC(=N1)C1=CC=CS1)N1C=CC(=N1)C1=CC=CS1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results