Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrotris[3-(2-thienyl)pyrazol-1-yl]borate, thallium salt, 95%, Thermo Scientific™

Catalog No. AC300150010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134030-71-0
Molecular Formula C21H16BN6S3Tl
Molecular Weight (g/mol) 663.77
MDL Number MFCD00800556
InChI Key CLSWVTDHYMQQPF-UHFFFAOYSA-N
PubChem CID 56845816
IUPAC Name thallium(1+);tris[(3-thiophen-2-ylpyrazol-1-yl)oxy]boron(1-)
SMILES [B-](ON1C=CC(=N1)C2=CC=CS2)(ON3C=CC(=N3)C4=CC=CS4)ON5C=CC(=N5)C6=CC=CS6.[Tl+]

Specifications

Melting Point 218.0°C to 220.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
Formula Weight 663.77
Percent Purity 95%
Physical Form Fine Crystalline Powder
Chemical Name or Material Hydrotris[3-(2-thienyl)pyrazol-1-yl]borate, thallium salt, 95%
missing translation for 'healthHazard1' GHS Signal Word: Danger
missing translation for 'healthHazard2' GHS H Statement
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
missing translation for 'healthHazard3' GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.