accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3SOONNHNHNF

CAS RN 1346547-00-9

IUPAC Name: N-(5-fluoro-1H-indazol-3-yl)-6-(2-methylpropane-2-sulfonyl)quinolin-4-amine
Synonyms: N-(5-fluoro-1H-indazol-3-yl)-6-(2-methylpropane-2-sulfonyl)quinolin-4-amine
Molecular Weight (g/mol): 398.46
Molecular Formula: C20H19FN4O2S
InChi Key: XLOGLWKOHPIJLV-UHFFFAOYSA-N
SMILES: CC(C)(C)S(=O)(=O)C1=CC=C2N=CC=C(NC3=NNC4=CC=C(F)C=C34)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)