accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1350555-93-9

IUPAC Name: N'-[4-(4-{[(4,5-diphenyl-1,3-oxazol-2-yl)sulfanyl]methyl}-6-[4-(hydroxymethyl)phenyl]-1,3-dioxan-2-yl)phenyl]-N-hydroxyoctanediamide
Synonyms: N'-[4-(4-{[(4,5-diphenyl-1,3-oxazol-2-yl)sulfanyl]methyl}-6-[4-(hydroxymethyl)phenyl]-1,3-dioxan-2-yl)phenyl]-N-hydroxyoctanediamide
Molecular Weight (g/mol): 721.87
Molecular Formula: C41H43N3O7S
InChi Key: BHUZLJOUHMBZQY-UHFFFAOYNA-N
SMILES: OCC1=CC=C(C=C1)C1CC(CSC2=NC(=C(O2)C2=CC=CC=C2)C2=CC=CC=C2)OC(O1)C1=CC=C(NC(=O)CCCCCCC(=O)NO)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results