CAS RN 135112-27-5
CAS RN 135112-27-5
IUPAC Name:
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms:
2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol):
325.36
Molecular Formula:
C19H19NO4
InChi Key:
XQIRYUNKLVPVRR-UHFFFAOYNA-N
SMILES:
CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
1
–
1
of
1
results
1
Fmoc-2-Abu-OH, Advanced ChemTech
CAS: 135112-27-5 Molecular Formula: C19H19NO4 Molecular Weight (g/mol): 325.36 InChI Key: XQIRYUNKLVPVRR-UHFFFAOYNA-N IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid SMILES: CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O