CAS RN 136152-65-3
IUPAC Name:
but-2-enedioic acid; dimethyl(2-{3-[1-(pyridin-2-yl)ethyl]-1H-inden-2-yl}ethyl)amine
Synonyms:
butenedioic acid; dimethindene
Molecular Weight (g/mol):
408.5
Molecular Formula:
C24H28N2O4
InChi Key:
SWECWXGUJQLXJF-UHFFFAOYNA-N
SMILES:
OC(=O)C=CC(O)=O.CC(C1=C(CCN(C)C)CC2=CC=CC=C12)C1=CC=CC=N1
1
–
1
of
1
results
1
R&D Systems™ (S)-(+)-Dimethindene maleate
CAS: 136152-65-3 Molecular Formula: C24H28N2O4