accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1364269-06-6

IUPAC Name: 4-(2-amino-5-{4-[(dimethylamino)methyl]thiophen-2-yl}pyridin-3-yl)-2-[(5,5,5-trifluoropent-3-en-2-yl)oxy]benzamide
Synonyms: 4-(2-amino-5-{4-[(dimethylamino)methyl]thiophen-2-yl}pyridin-3-yl)-2-[(5,5,5-trifluoropent-3-en-2-yl)oxy]benzamide
Molecular Weight (g/mol): 490.55
Molecular Formula: C24H25F3N4O2S
InChi Key: LLEOOSLUHRQCHZ-UHFFFAOYNA-N
SMILES: CC(OC1=C(C=CC(=C1)C1=C(N)N=CC(=C1)C1=CC(CN(C)C)=CS1)C(N)=O)C=CC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results