accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13698-16-3

H3COONClCl

CAS RN 13698-16-3

IUPAC Name: ethyl N,N-dichlorocarbamate
Synonyms: ethyl dichlorocarbamate
Molecular Weight (g/mol): 157.98
Molecular Formula: C3H5Cl2NO2
InChi Key: PCGGNOFZZFFFLU-UHFFFAOYSA-N
SMILES: CCOC(=O)N(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results