Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dichlorourethane 98%, Thermo Scientific Chemicals

Catalog No. AC161820250
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13698-16-3
Molecular Formula C3H5Cl2NO2
Molecular Weight (g/mol) 157.98
MDL Number MFCD00000523
InChI Key PCGGNOFZZFFFLU-UHFFFAOYSA-N
Synonym n,n-dichlorourethane, n,n-dichlorourethan, ethyl dichlorocarbamate, carbamic acid, dichloro-, ethyl ester, dichlorourethane, ethyl dichlorocarbamate #, n,n-dichlorocarbamic acid ethyl ester, carbamic acid,n,n-dichloro-, ethyl ester, n,n-dichlorourethane, technical grade, n,n-5-2-amino-5-2-methylpropyl-4-thiazolyl-2-furanyl phosphinylidene bis-,1,1-diethylesterl-al
PubChem CID 26195
IUPAC Name ethyl N,N-dichlorocarbamate
SMILES CCOC(=O)N(Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.3490g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Refractive Index 1.4590 to 1.4610 (20°C, 589nm)
Linear Formula Cl2NCO2C2H5
Quantity 25 g
Formula Weight 157.98
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material N, N-Dichlorourethane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Suspected of causing cancer.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.