accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13749-38-7

ClBrClClBr

CAS RN 13749-38-7

IUPAC Name: 1,2-dibromo-1,1,2-trichloroethane
Synonyms: 1,2-dibromo-1,1,2-trichloroethane
Molecular Weight (g/mol): 291.19
Molecular Formula: C2HBr2Cl3
InChi Key: VXKOVUFOVCJHBA-UHFFFAOYNA-N
SMILES: ClC(Br)C(Cl)(Cl)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results