Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dibromo-1,1,2-trichloroethane, 99%, Thermo Scientific™

Catalog No. AC279030250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC279030250 25 g Glass bottle
AC279030050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC279030250 Supplier Thermo Scientific Chemicals Supplier No. 279030250

Chemical Identifiers

CAS 13749-38-7
Molecular Formula C2HBr2Cl3
Molecular Weight (g/mol) 291.2
MDL Number MFCD00018841
InChI Key VXKOVUFOVCJHBA-UHFFFAOYSA-N
Synonym ethane,1,2-dibromo-1,1,2-trichloro, acmc-20ap6p, 1,2-dibromotrichloro ethane
PubChem CID 114562
IUPAC Name 1,2-dibromo-1,1,2-trichloroethane
SMILES C(C(Cl)(Cl)Br)(Cl)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5700 to 1.5720
Quantity 25 g
Formula Weight 291.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,2-Dibromo-1,1,2-trichloroethane, 99%
Show More Show Less
EINECSNumber 237-330-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.