accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 137944-39-9

CH3H2N(R)(R)NH2CH3FFFFFF

CAS RN 137944-39-9

IUPAC Name: methyl[(1R,2R)-2-(methylazaniumyl)-1,2-bis[3-(trifluoromethyl)phenyl]ethyl]azanium
Synonyms: methyl[(1R,2R)-2-(methylammonio)-1,2-bis[3-(trifluoromethyl)phenyl]ethyl]azanium
Molecular Weight (g/mol): 378.36
Molecular Formula: C18H20F6N2
InChi Key: SBGOGHODWUZHIY-HZPDHXFCSA-P
SMILES: C[NH2+][C@@H]([C@H]([NH2+]C)C1=CC=CC(=C1)C(F)(F)F)C1=CC=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results