Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2R)-(+)-N,N'-Dimethyl-1,2-bis[3-(trifluoromethyl)phenyl]-1,2-ethane diamine, 99%, Thermo Scientific™

Catalog No. AC294642500
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137944-39-9
Molecular Formula C18H18F6N2
Molecular Weight (g/mol) 376.33
MDL Number MFCD00075560
InChI Key SBGOGHODWUZHIY-HZPDHXFCSA-N
Synonym 1r,2r-+-n,n'-dimethyl-1,2-bis 3-trifluoromethyl phenyl ethylenediamine, 1r,2r-+-n,n'-dimethyl-1,2-bis 3-trifluoromethyl phenyl-1, 2-ethanediamine, methyl 1r,2r-2-methylamino-1,2-bis 3-trifluoromethyl phenyl ethyl amine, pubchem12633, r,r-metf-dpen, 1r,2r-n,n'-dimethyl-1,2-bis 3-trifluoromethyl phenyl ethane-1,2-diamine, 1r,2r-n,n'-dimethyl-1,2-bis 3-trifluoromethyl phenyl ethylenediamine, 1r,2r-+-n,n'-dimethyl-1,2-bis 3-trifluoromethyl phenyl-1,2-ethane diamine, 1r,2r-+-n,n'-dimethyl-1,2-bis 3-trifluoromethyl phenyl-1,2-ethanediamine
PubChem CID 2724996
IUPAC Name (1R,2R)-N,N'-dimethyl-1,2-bis[3-(trifluoromethyl)phenyl]ethane-1,2-diamine
SMILES CNC(C1=CC(=CC=C1)C(F)(F)F)C(C2=CC(=CC=C2)C(F)(F)F)NC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 113°C to 115°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min
Packaging Glass bottle
Quantity 250 mg
Specific Rotation Condition + 17.00 (25.00°C c=0.5,CHCl3)
Specific Rotation + 17
Formula Weight 376.33
Percent Purity 99%
Physical Form Powder
Chemical Name or Material (1R,2R)-(+)-N,N'-Dimethyl-1,2-bis[3-(trifluoromethyl)phenyl]-1,2-ethane diamine, 99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.