accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSOONOHNH3CCl(E)

CAS RN 139298-40-1

IUPAC Name: N-[2-({[(2E)-3-(4-chlorophenyl)prop-2-en-1-yl](methyl)amino}methyl)phenyl]-N-(2-hydroxyethyl)-4-methoxybenzene-1-sulfonamide
Synonyms: N-[2-({[(2E)-3-(4-chlorophenyl)prop-2-en-1-yl](methyl)amino}methyl)phenyl]-N-(2-hydroxyethyl)-4-methoxybenzenesulfonamide
Molecular Weight (g/mol): 501.04
Molecular Formula: C26H29ClN2O4S
InChi Key: LLLQTDSSHZREGW-AATRIKPKSA-N
SMILES: COC1=CC=C(C=C1)S(=O)(=O)N(CCO)C1=CC=CC=C1CN(C)C\C=C\C1=CC=C(Cl)C=C1
Form 
  • (1)
  • (2)
Product Type 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results