accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1394854-52-4

IUPAC Name: 3-{[2-(pyridin-3-yl)-6-(2,3,4,5-tetrahydro-1H-3-benzazepin-3-yl)pyrimidin-4-yl]amino}propanoate
Synonyms: 3-{[2-(pyridin-3-yl)-6-(1,2,4,5-tetrahydro-3-benzazepin-3-yl)pyrimidin-4-yl]amino}propanoate
Molecular Weight (g/mol): 388.45
Molecular Formula: C22H22N5O2
InChi Key: LJIFOCRGDDQFJF-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCNC1=NC(=NC(=C1)N1CCC2=CC=CC=C2CC1)C1=CN=CC=C1
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results