accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1404437-62-2

IUPAC Name: N-[2-(3-oxo-3,4-dihydroquinoxalin-2-yl)-4-(propan-2-yl)phenyl]thiophene-2-carboxamide
Synonyms: N-[4-isopropyl-2-(3-oxo-4H-quinoxalin-2-yl)phenyl]thiophene-2-carboxamide
Molecular Weight (g/mol): 389.47
Molecular Formula: C22H19N3O2S
InChi Key: HWOYIOLMBQSTQS-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(=C(NC(=O)C2=CC=CS2)C=C1)C1=NC2=CC=CC=C2NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results