accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNONNONH2ONH2

CAS RN 142557-61-7

IUPAC Name: 7-amino-1-cyclopropyl-3-ethyl-2,4-dioxo-1H,2H,3H,4H-pyrido[2,3-d]pyrimidine-6-carboxamide
Synonyms: 7-amino-1-cyclopropyl-3-ethyl-2,4-dioxopyrido[2,3-d]pyrimidine-6-carboxamide
Molecular Weight (g/mol): 289.3
Molecular Formula: C13H15N5O3
InChi Key: HWODCHXORCTEGU-UHFFFAOYSA-N
SMILES: CCN1C(=O)N(C2CC2)C2=NC(N)=C(C=C2C1=O)C(N)=O
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results