CAS RN 142715-48-8
IUPAC Name:
4-(6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-yl)-N-[2-(3,4-dimethoxyphenyl)ethyl]-6,7-dimethoxyquinazolin-2-amine hydrochloride
Synonyms:
4-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)-N-[2-(3,4-dimethoxyphenyl)ethyl]-6,7-dimethoxyquinazolin-2-amine hydrochloride
Molecular Weight (g/mol):
597.11
Molecular Formula:
C31H37ClN4O6
InChi Key:
WWCHXVYTCMPAMV-UHFFFAOYSA-N
SMILES:
Cl.COC1=CC2=NC(NCCC3=CC(OC)=C(OC)C=C3)=NC(N3CCC4=CC(OC)=C(OC)C=C4C3)=C2C=C1OC
1
–
1
of
1
results
1
R&D Systems™ CP 100356 hydrochloride
CAS: 142715-48-8 Molecular Formula: C31H37ClN4O6