CAS RN 1435934-00-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tocris Bioscience™ LDN 193189 in solution
Sterile-filtered 10 mM solution of LDN 193189 pre-dissolved in water
2
Tocris Bioscience™ LDN 193189
LDN 193189 synthesized to cGMP guidelines
3
Tocris Bioscience™ LDN 193189 dihydrochloride
Potent and selective ALK2 and ALK3 inhibitor; inhibits BMP4 signaling; promotes neural induction of hPSCs