CAS RN 144678-63-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ NSC 668036
Wnt signaling inhibitor; binds the PDZ domain of the Dishevelled (Dvl) protein
2
Tocris Bioscience™ Ex 26
Potent and selective S1P1 receptor antagonist