accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH2SNNHClHBrHBr(E/Z)

CAS RN 145231-35-2

IUPAC Name: N'-[(4-chlorophenyl)methyl]{[3-(1H-imidazol-5-yl)propyl]sulfanyl}methanimidamide dihydrobromide
Synonyms: clobenpropit dihydrobromide
Molecular Weight (g/mol): 470.65
Molecular Formula: C14H19Br2ClN4S
InChi Key: JIJQPEZAVLJZBO-UHFFFAOYSA-N
SMILES: Br.Br.NC(SCCCC1=CN=CN1)=NCC1=CC=C(Cl)C=C1
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results