CAS RN 145307-34-2
IUPAC Name:
hydrogen (1R)-3-(adamantan-1-yl)-1-(aminomethyl)-3,4-dihydro-1H-2-benzopyran-5,6-diol chloride
Synonyms:
hydrogen (1R)-3-(adamantan-1-yl)-1-(aminomethyl)-3,4-dihydro-1H-2-benzopyran-5,6-diol chloride
Molecular Weight (g/mol):
365.9
Molecular Formula:
C20H28ClNO3
InChi Key:
BWHPNJVKFAPVOG-LXJUREGMSA-N
SMILES:
[H+].[Cl-].NC[C@@H]1OC(CC2=C(O)C(O)=CC=C12)C12CC3CC(CC(C3)C1)C2
1
–
1
of
1
results
1
R&D Systems™ A 77636 hydrochloride
CAS: 145307-34-2 Molecular Formula: C20H28ClNO3