CAS RN 1480833-70-2
CAS RN 1480833-70-2
IUPAC Name:
                                            
                                                    2-{5-[4-(trifluoromethoxy)phenyl]-1H-imidazol-2-yl}propan-2-amine
                                            
                                        
                                    
                                    
                                    
                                    
                                        Synonyms:
                                        
                                                2-{4-[4-(trifluoromethoxy)phenyl]-3H-imidazol-2-yl}propan-2-amine
                                        
                                    
                                                Molecular Weight (g/mol):
                                                
                                                           285.27
                                                
                                            
                                            
                                                 Molecular Formula:
                                                
                                                        C13H14F3N3O
                                            
                                                
                                            
                                        InChi Key:
                                                
                                                        AGORGFNWYAUYSU-UHFFFAOYSA-N
                                                
                                            
                                            
                                            SMILES:
                                                
                                                            CC(C)(N)C1=NC=C(N1)C1=CC=C(OC(F)(F)F)C=C1
                                                
                                            
                                        
                                        1
                                        –
                                        1
                                        of
                                        1
                                        results
                                    
                                
                                        1 
                                    
                            
                                    
                                                        Tocris Bioscience™ PF 04885614
                                                
                                                
                                                
                                                    
                                                
                                                            
                                                             
                                                                
                                                            
                                                    
                                                
                                                                         
                                                                
                                                            
                                                    CAS: 1480833-70-2 Molecular Formula: C13H14F3N3O
