CAS RN 149013-84-3
CAS RN 149013-84-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific Chemicals Trimethylstannylmethyldiphenylsilan, 96%, Thermo Scientific™
CAS: 149013-84-3 Molecular Formula: C16H22SiSn Molecular Weight (g/mol): 360.93 MDL Number: MFCD00800634 Synonym: silane,diphenyl trimethylstannyl methyl-9ci,acmc-1cjkr