Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trimethylstannylmethyldiphenylsilan, 96%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Specifications
Specifications
Color | Colorless |
Boiling Point | 125°C to 130°C |
Assay Percent Range | 96% |
Packaging | Glass bottle |
Linear Formula | (CH3)3SNSI(C6H5)2CH3 |
Quantity | 1 g |
Formula Weight | 360.93 |
Percent Purity | 96% |
Physical Form | Liquid |
Chemical Name or Material | Trimethylstannylmethyldiphenylsilan, 96% |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Fatal if inhaled. Causes damage to organs through prolonged or repeated exposure. Causes skin irritation. Fatal in contact with skin. Causes serious eye irritation. Fatal if swallowed. Very toxic |
Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction