CAS RN 149409-57-4
IUPAC Name:
hydrogen diethyl({2-[4-methoxy-3-(2-phenylethoxy)phenyl]ethyl})amine chloride
Synonyms:
hydrogen diethyl({2-[4-methoxy-3-(2-phenylethoxy)phenyl]ethyl})amine chloride
Molecular Weight (g/mol):
363.93
Molecular Formula:
C21H30ClNO2
InChi Key:
LPMIMPZUXJWRJJ-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CCN(CC)CCC1=CC=C(OC)C(OCCC2=CC=CC=C2)=C1
1
–
1
of
1
results
1
R&D Systems™ NE 100 hydrochloride
CAS: 149409-57-4 Molecular Formula: C23H34ClNO2