accessibility menu, dialog, popup

UserName

Viewing profile as user:
CHNNNHONH2NHN

CAS RN 1507367-00-1

IUPAC Name: 2-{3-[(6-ethynyl-7H-purin-2-yl)amino]phenyl}acetamide
Synonyms: 2-{3-[(6-ethynyl-7H-purin-2-yl)amino]phenyl}acetamide
Molecular Weight (g/mol): 292.3
Molecular Formula: C15H12N6O
InChi Key: CJQGLUJHMXCLQY-UHFFFAOYSA-N
SMILES: NC(=O)CC1=CC(NC2=NC(C#C)=C3NC=NC3=N2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)