accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONSClNNNOO(E)

CAS RN 153719-23-4

IUPAC Name: (4E)-3-[(2-chloro-1,3-thiazol-5-yl)methyl]-5-methyl-N-nitro-1,3,5-oxadiazinan-4-imine
Synonyms: C8H10clN5O3S (E)-thiamethoxam
Molecular Weight (g/mol): 291.71
Molecular Formula: C8H10ClN5O3S
InChi Key: NWWZPOKUUAIXIW-DHZHZOJOSA-N
SMILES: CN1COCN(CC2=CN=C(Cl)S2)\C1=N\[N+]([O-])=O
Color 
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)