accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SN(S)(S)OOOCH3(S)H3CONSH3CHNO

CAS RN 156177-65-0

IUPAC Name: methyl (15S,16S,18S)-10,23-bis[(ethylsulfanyl)methyl]-15-methyl-3-oxo-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8(13),9,11,20,22,24-nonaene-16-carboperoxoate
Synonyms: methyl (15S,16S,18S)-10,23-bis[(ethylsulfanyl)methyl]-15-methyl-3-oxo-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8(13),9,11,20,22,24-nonaene-16-carboperoxoate
Molecular Weight (g/mol): 615.76
Molecular Formula: C33H33N3O5S2
InChi Key: WMBUVOLZSWTKMP-KQFGJPIXSA-N
SMILES: CCSCC1=CC=C2N3[C@@H]4C[C@H](C(=O)OOC)[C@](C)(O4)N4C5=C(C=C(CSCC)C=C5)C5=C4C3=C(C2=C1)C1=C5CNC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results