accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 156939-62-7

OOONH

CAS RN 156939-62-7

IUPAC Name: (9H-fluoren-9-yl)methyl N-(2-oxoethyl)carbamate
Synonyms: 9H-fluoren-9-ylmethyl N-(2-oxoethyl)carbamate
Molecular Weight (g/mol): 281.31
Molecular Formula: C17H15NO3
InChi Key: DBTMQODRSDEGRZ-UHFFFAOYSA-N
SMILES: O=CCNC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)