CAS RN 15960-05-1
CAS RN 15960-05-1
IUPAC Name:
(2S)-2-azaniumyl-4,4,4-trifluorobutanoate
Synonyms:
(2S)-2-ammonio-4,4,4-trifluorobutanoate
Molecular Weight (g/mol):
157.09
Molecular Formula:
C4H6F3NO2
InChi Key:
AQPCXCOPDSEKQT-REOHCLBHSA-N
SMILES:
[NH3+][C@@H](CC(F)(F)F)C([O-])=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals 2-Amino-4,4,4-trifluorobutyric acid, 97%, Thermo Scientific™
CAS: 15960-05-1 Molecular Formula: C4H6F3NO2 Molecular Weight (g/mol): 157.09 MDL Number: MFCD17676355 InChI Key: AQPCXCOPDSEKQT-REOHCLBHSA-N Synonym: 2s-2-amino-4,4,4-trifluorobutanoic acid,s-2-amino-4,4,4-trifluorobutanoic acid,s-2-amino-4,4,4-trifluorobutyric acid,4,4,4-trifluoro-l-abu-oh,2s-2-amino-4,4,4-trifluorobu-tanoic acid PubChem CID: 6993161 IUPAC Name: (2S)-2-amino-4,4,4-trifluorobutanoic acid SMILES: [NH3+][C@@H](CC(F)(F)F)C([O-])=O