Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4,4,4-trifluorobutyric acid, 97%, Thermo Scientific™

Catalog No. AC297352500
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15960-05-1
Molecular Formula C4H6F3NO2
Molecular Weight (g/mol) 157.09
MDL Number MFCD17676355
InChI Key AQPCXCOPDSEKQT-REOHCLBHSA-N
Synonym 2s-2-amino-4,4,4-trifluorobutanoic acid, s-2-amino-4,4,4-trifluorobutanoic acid, s-2-amino-4,4,4-trifluorobutyric acid, 4,4,4-trifluoro-l-abu-oh, 2s-2-amino-4,4,4-trifluorobu-tanoic acid
PubChem CID 6993161
IUPAC Name (2S)-2-amino-4,4,4-trifluorobutanoic acid
SMILES [NH3+][C@@H](CC(F)(F)F)C([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 256°C to 258°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CF3CH2CH(NH2)COOH
Quantity 250 mg
Formula Weight 157.09
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4, 4, 4-trifluorobutyric acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.