accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNH2ONH(R)OONNSCH3OOCH3SOHOO

CAS RN 159752-10-0

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)