accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16142-27-1

ClNH2NNONO

CAS RN 16142-27-1

IUPAC Name: 5-amino-3-(morpholin-4-yl)-3H-1λ⁴,2,3-oxadiazol-1-ylium chloride
Synonyms: 5-amino-3-(morpholin-4-yl)-1λ⁴,2,3-oxadiazol-1-ylium chloride
Molecular Weight (g/mol): 206.63
Molecular Formula: C6H11ClN4O2
InChi Key: ZRFWHHCXSSACAW-UHFFFAOYSA-M
SMILES: [Cl-].NC1=CN(N=[O+]1)N1CCOCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)