CAS RN 1627709-94-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ LJI308
Potent pan-ribosomal S6 protein kinase (RSK) inhibitor (IC50 values are 4 to 13 nM)