CAS RN 1642187-96-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ TASP 0390325
Potent and selective V1B receptor antagonist; orally bioavailable