CAS RN 169132-80-3
CAS RN 169132-80-3
IUPAC Name:
O,O,O-tris[4-({2-[bis(4-formylphenoxy)(sulfanylidene)-λ⁵-phosphanyl]-2-methylhydrazin-1-ylidene}methyl)phenyl] phosphorothioate
Synonyms:
O,O,O-tris[4-({2-[bis(4-formylphenoxy)(sulfanylidene)-λ⁵-phosphanyl]-2-methylhydrazin-1-ylidene}methyl)phenyl] phosphorothioate
Molecular Weight (g/mol):
1423.32
Molecular Formula:
C66H54N6O15P4S4
InChi Key:
GVUPCQKZNKROEB-UHFFFAOYSA-N
SMILES:
CN(N=CC1=CC=C(OP(=S)(OC2=CC=C(C=NN(C)P(=S)(OC3=CC=C(C=O)C=C3)OC3=CC=C(C=O)C=C3)C=C2)OC2=CC=C(C=NN(C)P(=S)(OC3=CC=C(C=O)C=C3)OC3=CC=C(C=O)C=C3)C=C2)C=C1)P(=S)(OC1=CC=C(C=O)C=C1)OC1=CC=C(C=O)C=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Thiophosphoryl-PmmH-6 Dendrimer, Generation 1.5, Thermo Scientific™
CAS: 169132-80-3 Molecular Formula: C66H54N6O15P4S4 Molecular Weight (g/mol): 1423.32 MDL Number: MFCD03458724 InChI Key: GVUPCQKZNKROEB-UHFFFAOYSA-N Synonym: thiophosphoryl-pmmh-6 dendrimer, generation 1.5 PubChem CID: 131874880 SMILES: CN(N=CC1=CC=C(C=C1)OP(=S)(OC2=CC=C(C=C2)C=NN(C)P(=S)(OC3=CC=C(C=C3)C=O)OC4=CC=C(C=C4)C=O)OC5=CC=C(C=C5)C=NN(C)P(=S)(OC6=CC=C(C=C6)C=O)OC7=CC=C(C=C7)C=O)P(=S)(OC8=CC=C(C=C8)C=O)OC9=CC=C(C=C9)C=O