accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOHO

CAS RN 170355-78-9

IUPAC Name: 4-[7-(adamantan-1-yl)-6-[(2-methoxyethoxy)methoxy]naphthalen-2-yl]benzoic acid
Synonyms: 4-[7-(adamantan-1-yl)-6-[(2-methoxyethoxy)methoxy]naphthalen-2-yl]benzoic acid
Molecular Weight (g/mol): 486.61
Molecular Formula: C31H34O5
InChi Key: JBALRFFXKQPVLT-UHFFFAOYSA-N
SMILES: COCCOCOC1=C(C=C2C=C(C=CC2=C1)C1=CC=C(C=C1)C(O)=O)C12CC3CC(CC(C3)C1)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results