accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNClHN

CAS RN 178946-89-9

IUPAC Name: 3-[(4-chlorophenyl)(1H-indol-3-yl)methyl]-1H-indole
Synonyms: 3-[(4-chlorophenyl)(1H-indol-3-yl)methyl]-1H-indole
Molecular Weight (g/mol): 356.85
Molecular Formula: C23H17ClN2
InChi Key: LTLRXTDMXOFBDW-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(C1=CNC2=CC=CC=C12)C1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results